Anti-AADAT Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-416EA 480 EUR
PRSI48-416
Anti-AADAT Rabbit Polyclonal Antibody
Kynurenine aminotransferase catalyzes the irreversible transamination of the L-tryptophan metabolite L-kynurenine to form kynurenic acid. Kynurenic acid is an antagonist at the glycine site of NMDA receptors and increased levels of kynurenic acid are thought to exert a neuroprotective action in some pathological conditions.

AADAT antibody can be used in Western Blot starting at 0.5 - 2 μg/mL, immunohistochemistry starting at 10 μg/mL, and immunoprecipitation.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: AADAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR