Anti-PLA2G3 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-414EA 480 EUR
PRSI48-414
Anti-PLA2G3 Rabbit Polyclonal Antibody
Antikörper
Human group III secreted phospholipase A2 belongs to a superfamily of intracellular and secreted enzymes that catalyze the hydrolysis of glycerophospholipids at the sn-2 position to release fatty acids and lysophospholipids. Group III PLA2 enzymes were originally identified in invertebrates such as bees and scorpions. Human group III sPLA2 has been shown to be a calcium dependent enzyme. It consists of a central group III sPLA2 domain flanked by unique N- and C-terminal domains that are proteolytically removed in most tissues. The sPLA2 domain is sufficient for catalytic activity. SPLA2-III is preferentially expressed in the microvascular endothelium of tissues with inflammation, ischemic injury and cancer.

PLA2G3 antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:200 - 1:400.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PLA2G3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR