Anti-MMP9 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-413EA 480 EUR
PRSI48-413
Anti-MMP9 Rabbit Polyclonal Antibody
Antikörper
Matrix metalloproteinases (MMPs) are neutral proteinases that are involved in the breakdown and remodeling of the extracellular matrix (ECM) under a variety of physiological and pathological conditions. MMP9 is a Metallopeptidase M10A type protease and is particularly important in the pathogenesis of inflammatory, infectious, and neoplastic diseases in many organs.

MMP9 antibody can be used in ELISA starting at 1:10000, and immunohistochemistry starting at 1:100 - 1:200.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR