Anti-KDR Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-411EA 480 EUR
PRSI48-411
Anti-KDR Rabbit Polyclonal Antibody
Antikörper
KDR is a VEGF Receptor type protein kinase.

KDR antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:200.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KDR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR