Anti-KDR Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-410EA 619 EUR
PRSI48-410
Anti-KDR Rabbit Polyclonal Antibody
Antikörper
KDR is a VEGF Receptor type protein kinase.

KDR antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KDR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Dog, Gorilla, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR