Anti-FLT1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-407EA 480 EUR
PRSI48-407
Anti-FLT1 Rabbit Polyclonal Antibody
Antikörper
FLT1 (fms-like tyrosine kinase/vascular endothelial growth factor receptor 1) is a receptor for VEGR, VEGFB, and PGF. This receptor has a large extracellular domain similar to other members of the fms family and has tyrosine kinase activity. The VEGF kinase ligand/receptor signaling system plays a key role in vascular development and regulation of vascular permeability.

FLT1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FLT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR