Anti-INSR Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-403EA 619 EUR
PRSI48-403
Anti-INSR Rabbit Polyclonal Antibody
Antikörper
INSR is an insulin receptor type protein kinase. Binding of insulin to INSR stimulates glucose uptake. Two insulin receptor mRNA transcripts resulting from alternative splicing of exon 11 in the receptor gene are expressed in a highly regulated tissue-specific fashion. Mutations in the INSR gene contribute to diseases, including of various forms of insulin resistance, insulin-resistant diabetes mellitus, noninsulin-dependent diabetes mellitus (NIDDM), Rabson-Mendenhall syndrome and Leprechaunism (Donohue Syndrome).

Insulin Receptor antibody can be used in ELISA starting at 1:10000, and immunohistochemistry starting at 1:200 - 1:400.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: INSR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR