Anti-ITGB1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-392EA 480 EUR
PRSI48-392
Anti-ITGB1 Rabbit Polyclonal Antibody
Antikörper
Integrins are a major class of cell adhesion molecules that mediate cell matrix interactions. They are composed of an alpha and a beta subunit. Beta 1 heterodimers are widely expressed and control various developmental processes, including neurogenesis, chondrogenesis, myoblast fusion, and skin and hair follicle morphogenesis.

Integrin beta 1 antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:200.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ITGB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey, Orangutan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR