Anti-ITGAX Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-386EA 480 EUR
PRSI48-386
Anti-ITGAX Rabbit Polyclonal Antibody
Antikörper
ITGAX encodes the integrin alpha X chain. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This I-domain containing alpha integrin combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 seems to broadly overlap the properties of the alpha M beta 2 integrin in adherence of neutrophils and monocytes to stimulated endothelium, and the phagocytosis of complement coated particles.

Integrin alpha X antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, and immunoprecipitation

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ITGAX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR