Anti-TRPA1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-380EA 480 EUR
PRSI48-380
Anti-TRPA1 Rabbit Polyclonal Antibody
Antikörper
TRPA1 has been suggested to be a cold-activated ion channel. It has also been suggested to affect mechanotransduction and the perception of sound in vertebrates.

TRPA1 antibody can be used in Western Blot starting at 2 μg/mL, immunohistochemistry starting at 10 μg/mL, immunoprecipitation starting at 2 μg/mL, and flow cytometry starting at 2 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR