Anti-USP33 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-379EA 480 EUR
PRSI48-379
Anti-USP33 Rabbit Polyclonal Antibody
VDU1 (pVHL-interacting deubiquitinating enzyme-1), is a substrate of von Hippel-Lindau tumor suppressor protein (pVHL) E3 ligase complex. VDU1 contains the signature motifs of the ubiquitin-specific processing protease family and possesses deubiquitinating activity. VDU1 and VDU2 define a subfamily of ubiquitin specific processing proteases.

USP33 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: USP33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Dog, Gibbon, Gorilla, Horse, Human, Orangutan, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR