Anti-WNT10A Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-377EA 480 EUR
PRSI48-377
Anti-WNT10A Rabbit Polyclonal Antibody
Antikörper
Wnts are important signaling molecules involved in normal developmental processes as well as some forms of cancer. WNT10A expression has been documented in many cancers, include those of the stomach, colon, breast, bone marrow, esophagus, and blood.

WNT10A antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200 - 1:400, and flow cytometry starting at 1:200 - 1:400.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: WNT10A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Dog, Gibbon, Gorilla, Human, Monkey, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR