Anti-EPHX2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-376EA 480 EUR
PRSI48-376
Anti-EPHX2 Rabbit Polyclonal Antibody
Antikörper
Soluble Epoxide Hydrolase (sEH) also known as cytosolic Epoxide Hydrolase is a phase I xenobiotic-metabolizing enzyme that converts epoxyeicosatrienoic acids (EETs) into their corresponding dihydroxyeicosatrienoic acids (DHETs). EETs are thought to be involved in vascular tone, ion channel regulation, mitogenesis and cell signaling. DHETs have, in general, less biological activity then EETs and are more readily excreted, thus hydration of EETs by sEH maybe a means of attenuating or eliminating their activity.sEHs have been identified in many tissues including liver, kidney, adrenal, pancreatic islets, gi tract and vascular smooth muscle.

EPHX2 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunoprecipitation

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EPHX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR