Anti-PTGES Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-375EA 480 EUR
PRSI48-375
Anti-PTGES Rabbit Polyclonal Antibody
Antikörper
PGES-1 is a membrane associated protein involved in eicosanoid and glutathione metabolism as part of the arachidonic acid signaling pathway. The expression of this protein is induced by the proinflammatory cytokine IL-1B and its’ activity is glutathione dependent. PGES is also expressed at high levels in P53 expressing cells.

PTGES antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 1:200 - 1:400.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTGES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Chicken, Dog, Gorilla, Horse, Human, Monkey, Pig, Rabbit, Rat, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR