Anti-WNT5B Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-370EA 619 EUR
PRSI48-370
Anti-WNT5B Rabbit Polyclonal Antibody
Antikörper
WNT signaling pathway plays key roles in carcinogenesis and embryogenesis, and WNT signaling molecules are potent targets for diagnosis, prevention and treatment of cancer as well as for regenerative medicine or tissue engineering.

WNT5B antibody can be used in immunohistochemistry starting at 1:50.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: WNT5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Human, Orangutan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR