Anti-CX3CR1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-367EA 480 EUR
PRSI48-367
Anti-CX3CR1 Rabbit Polyclonal Antibody
Antikörper
CX3CR1 a Chemokine Receptor is an HIV coreceptor, as well as a leukocyte chemotactic/adhesion receptor for fractalkine, also known as neurotactin. CX3CR1 mediates chemotactic responses to the soluble form of fractalkine in neutrophils and T cells, and cytoadhesion to cells expressing membrane-anchored fractalkine, such as monocytes, T cells, and neutrophils. Cells transfected with CX3CR1 exhibited chemotaxis and transient increases in intracellular calcium concentration when exposed to fractalkine.

CX3CR1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CX3CR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR