Anti-WNT8A Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-357EA 480 EUR
PRSI48-357
Anti-WNT8A Rabbit Polyclonal Antibody
Antikörper
WNT8A antibody can be used in immunohistochemistry starting at 10 μg/mL, and immunofluorescence.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: WNT8A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Gorilla, Horse, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR