Anti-PRKCZ Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-355EA 480 EUR
PRSI48-355
Anti-PRKCZ Rabbit Polyclonal Antibody
Antikörper
Protein Kinase C zeta antibody can be used in ELISA, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PRKCZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR