Anti-ATRN Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-352EA 480 EUR
PRSI48-352
Anti-ATRN Rabbit Polyclonal Antibody
Antikörper
Attractin antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ATRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Dog, Gibbon, Hamster, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR