Anti-FAP Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-343EA 480 EUR
PRSI48-343
Anti-FAP Rabbit Polyclonal Antibody
Antikörper
FAP antibody can be used in Western Blot, immunohistochemistry starting at 10 μg/mL, and immunoprecipitation.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR