Anti-HTR7 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-340EA 619 EUR
PRSI48-340
Anti-HTR7 Rabbit Polyclonal Antibody
Antikörper
5-HT7 Receptor is a Serotonin Receptor with several functions, including regulation of the circadian rhythm and mediation of smooth-muscle relaxation in the vasculature and GI tract. This receptor has been implicated in depression, autistism, and other neuropsychiatric disorders. Three protein isoforms have been documented: isoform a: 445aa; isoform b: 432aa; isoform d: 479aa.

HTR7 antibody can be used in ELISA, and immunohistochemistry starting at 20 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HTR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Dog, Gibbon, Gorilla, Guinea pig, Hamster, Horse, Human, Monkey, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR