Anti-PCSK6 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-339EA 480 EUR
PRSI48-339
Anti-PCSK6 Rabbit Polyclonal Antibody
PACE4 is a Serine S8 type protease that cleaves precursor proteins at paired basic amino acid processing sites. Several PACE4 substrates have been identified, including transforming growth factor beta-related proteins, proalbumin and von Willebrand factor. At least eight alternatively spliced transcript variants encoding different isoforms have been reported. In mouse, PACE4 has been shown to increase tumor cell invasiveness, and thus enhance tumor progression.

PCSK6 antibody can be used in ELISA, and immunohistochemistry starting at 20 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PCSK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR