Anti-WNK1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-335EA 480 EUR
PRSI48-335
Anti-WNK1 Rabbit Polyclonal Antibody
Antikörper
WNK1, a WNK type protein kinase, contains a small N-terminal domain followed by a kinase domain and a long C-terminal tail. The protein is cytoplasmic and has been shown to regulate both the c-Jun N-terminal kinase pathway and the actin cytoskeleton in vitro. WNK1 has been identified as the gene mutant in pseudohypoaldosteronism type 2, also known as Gordon hyperkalemia-hypertension syndrome, which is a rare autosomal dominant form of volume-dependent, low-renin hypertension. Disease-causing mutations are large intronic deletions that increase WNK1 expression. At least two variants of WNK1 are produced by alternative splicing.

WNK1 antibody can be used in immunohistochemistry starting at 6 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: WNK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Chicken, Gibbon, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR