Anti-PDE2A Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-327EA 480 EUR
PRSI48-327
Anti-PDE2A Rabbit Polyclonal Antibody
Phosphodiesterase 2A is a cGMP stimulated, cAMP/cGMP dual-specific phosphodiesterase whose hydrolytic activity is stimulated by the presence of cGMP binding to the GAF domain. PDE2A has been implicated in penile erectile dysfunction and in the regulation of fluid and inflammatory cell transit across the endothelial cell barrier (i.e., venous and capillary). PDE2A activity is inhibited by erythro-9-(2-hydroxyl-3-nonyl)adenine (EHNA). At least three isoforms have been reported (PDE2A1, PDE2A2, and PDE2A3).

PDE2A antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDE2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Gorilla, Horse, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR