Anti-PDE3A Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-326EA 480 EUR
PRSI48-326
Anti-PDE3A Rabbit Polyclonal Antibody
Antikörper
Phosphodiesterase 3A is a cGMP inhibited, cAMP/cGMP dual-specific phosphodiesterase. PDE3A has been implicated in male erectile dysfunction, mammalian oocyte maturation, platelet function, vascular smooth muscle function, and salivary gland tumors. Three alternatively spliced transcripts have been reported.

PDE3A antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PDE3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR