Anti-MRGPRX2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-297EA 514 EUR
PRSI48-297
Anti-MRGPRX2 Rabbit Polyclonal Antibody
MRGX2 is highly similar to other MRG receptors. Recently it has been shown that MrgX2 is a high potency cortistatin receptor expressed in dorsal root ganglion.

MRGPRX2 antibody can be used in immunohistochemistry starting at 20 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MRGPRX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR