Anti-FFAR2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-293EA 480 EUR
PRSI48-293
Anti-FFAR2 Rabbit Polyclonal Antibody
Antikörper
GPR43 is classified as a Carboxylic Acid GPCR. The location of this receptor in the immune system is suggestive of potential roles in immune cell function and hematopoiesis.

FFAR2 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 2.5 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FFAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR