Anti-FFAR2 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-292EA 619 EUR
PRSI48-292
Anti-FFAR2 Rabbit Polyclonal Antibody
Antikörper
GPR43 is classified as a Carboxylic Acid GPCR. The location of this receptor in the immune system is suggestive of potential roles in immune cell function and hematopoiesis.

FFAR2 antibody can be used in immunohistochemistry starting at 7 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FFAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR