Anti-HTR1E Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-290EA 480 EUR
PRSI48-290
Anti-HTR1E Rabbit Polyclonal Antibody
Antikörper
5HT1E, a Serotonin Receptor, is not well studied. It is classified as an 5-HT1-like receptor because of its ligand-binding and effector-coupling characteristics. The human 5-HT1E receptor has been studied in an African green monkey kidney cell line, in which it modulated cAMP levels upon binding serotonin. Lack of structural variants in the human gene indicate a highly conserved protein.

HTR1E antibody can be used in immunohistochemistry starting at 10 - 15 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HTR1E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Chimpanzee, Guinea pig, Gorilla, Horse, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR