Anti-GPR88 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-289EA 480 EUR
PRSI48-289
Anti-GPR88 Rabbit Polyclonal Antibody
Antikörper
GPR88 is an Orphan-A GPCR with an unknown ligand.

GPR88 antibody can be used in immunohistochemistry starting at 6 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR88
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Gorilla, Hamster, Human, Monkey, Mouse, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR