Anti-RARB Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-285EA 480 EUR
PRSI48-285
Anti-RARB Rabbit Polyclonal Antibody
Antikörper
Retinoic acid receptor beta (RAR beta), a NR1 Thyroid Hormone-Like Receptor, is involved in the regulation of cell growth, differentiation, apoptosis, and carcinogenesis. RAR beta forms heterodimers with the retinoic acid X receptor (RXR) and binds to the direct repeat 1 (DR1) sequence of target DNA. A growing body of evidence supports the hypothesis that RAR beta is a tumor-suppressor gene. Altered expression of RAR beta has been linked to cancers, including those of the head and neck, lung, esophagus, breast, pancreas, and cervix. Three isoforms of RAR beta, beta 1, beta 2, and beta 4, have been documented in humans.

Retinoic Acid Receptor beta antibody can be used in immunohistochemistry starting at 10 - 15 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RARB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Gibbon, Gorilla, Horse, Human, Monkey, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR