Anti-NR0B1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-283EA 480 EUR
PRSI48-283
Anti-NR0B1 Rabbit Polyclonal Antibody
Antikörper
DAX-1 (DSS-AHC critical region on the X chromosome, gene 1) is an orphan NRO Knirps-Like nuclear receptor that is involved in adrenal gland and gonad development. DAX-1 has been shown to negatively regulate transcription of the nuclear receptor steroidogenic factor 1 (SF-1). DAX-1 is also involved in the development and differentiation of anterior pituitary cells and in steroidogenesis. In the nucleus, DAX-1 binds to a retinoic acid-responsive element (RARE) and acts as a dominant-negative regulator of retinoic acid receptor-mediated transcription. DAX-1 gene transcription is regulated by Ad4BP/SF-1 (NR5A1) and is suppressed by androgens and the androgen receptor (AR/NR3C4); AR down-regulated DAX-1 gene transcription is mediated by Ad4BP/SF-1 in the presence of the ligand. Recently it has been shown that DAX-1 is required for testis development (Meeks et al. 2003). Mutations in the human DAX-1 gene lead to X-linked adrenal hypoplasia congenita and hypogonadotropic hypogonadism. Recently two isoforms of DAX-1 has been identified.

NR0B1 antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 1:200 - 1:400, and immunofluorescence starting at 1:200 - 1:1000.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NR0B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Chimpanzee, Gorilla, Human, Orangutan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR