Anti-GPR160 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-281EA 480 EUR
PRSI48-281
Anti-GPR160 Rabbit Polyclonal Antibody
Antikörper
GPR160/GPCR150 is an Orphan-A GPCR with an unknown ligand.

GPR160 antibody can be used in immunohistochemistry starting at 6 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR160
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR