Anti-NR5A1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-278EA 619 EUR
PRSI48-278
Anti-NR5A1 Rabbit Polyclonal Antibody
Antikörper
Steroidogenic factor 1 (SF-1), a NR5 Fushi Tarazu-Like Receptor, has been shown to affect sexual differentiation, steroid metabolism, and neural development. SF-1 binds as a monomer and regulates all steroidogenic p450 genes, Mullerian inhibiting substance, the gonadotrophins, the ACTH receptor, oxytocin, nuclear receptor DAX-1, and several other genes. Four alternatively spliced isoforms of SF-1 have been isolated in mice, and they differ at the N and C termini but are identical in the DNA-binding domain.

Steroidogenic Factor 1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NR5A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR