Anti-GPR149 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-277EA 480 EUR
PRSI48-277
Anti-GPR149 Rabbit Polyclonal Antibody
Antikörper
The G protein-coupled receptor R35 is an Orphan-U GPCR with an unknown ligand.

GPR149 antibody can be used in immunohistochemistry starting at 4 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR149
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR