Anti-FZD9 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-275EA 480 EUR
PRSI48-275
Anti-FZD9 Rabbit Polyclonal Antibody
Antikörper
FZD9, previously named frizzled-3, is a Frizzled Receptor that mediates Wnt signaling. It may contribute to the Williams-Beuren syndrome described as supravalvular and aortic stenosis, and to infantile hypercalcemia.

Frizzled 9 antibody can be used in immunohistochemistry starting at 15 - 20 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FZD9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR