Anti-GPR151 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-268EA 480 EUR
PRSI48-268
Anti-GPR151 Rabbit Polyclonal Antibody
Antikörper
TAAR5 is classified as an Orphan-A GPCR because the ligand has not been identified.

GPR151 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR151
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR