Anti-GPR37 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-265EA 480 EUR
PRSI48-265
Anti-GPR37 Rabbit Polyclonal Antibody
Antikörper
GPR37 is an Orphan-A GPCR with an unknown ligand. GPR37 was recently identified as the PAEL receptor, a Parkin substrate involved in autosomal recessive juvenile Parkinson's (PDJ) disease. The PAEL receptor becomes unfolded, insoluble, and ubiquitinated when overexpressed, leading to unfolded protein-induced cell death. When the PAEL receptor is ubiquitinated by Parkin, it gets degraded, resulting in the suppression of cell death. The insoluble form of the PAEL receptor accumulates in the brains of PDJ patients and may cause selective neuronal death.

PAEL Receptor antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR37
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR