Anti-GPR52 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-263EA 480 EUR
PRSI48-263
Anti-GPR52 Rabbit Polyclonal Antibody
Antikörper
GPR52 is an Orphan-A GPCR with an unknown ligand. It has been identified from the high-throughput genome (HTG) database and subsequently cloned. It is localized to human chromosome 1.

GPR52 antibody can be used in immunohistochemistry starting at 15 - 30 μg/mL.

PBS, 0.1% sodium azide.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR