Anti-MRGPRF Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-141EA 480 EUR
PRSI48-141
Anti-MRGPRF Rabbit Polyclonal Antibody
MAS-related GPR, member F (MRGPRF) is an Orphan-U GPCR with an unknown ligand.

MRGPRF antibody can be used in immunohistochemistry starting at 10 μg/mL, immunoprecipitation, and flow cytometry.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MRGPRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR