Anti-OXGR1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-140EA 619 EUR
PRSI48-140
Anti-OXGR1 Rabbit Polyclonal Antibody
Antikörper
The G protein-coupled receptor GPR80/GPR99/P2Y15 is similar to the purinergic P2Y receptors.

OXGR1 antibody can be used in immunohistochemistry starting at 7 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: OXGR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR