Anti-GPR101 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-138EA 480 EUR
PRSI48-138
Anti-GPR101 Rabbit Polyclonal Antibody
Antikörper
G protein-coupled receptor GPR101 is an Orphan-U GPCR with an unknown ligand.

GPR101 antibody can be used in Western Blot, immunohistochemistry starting at 1:50, and flow cytometry.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR101
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR