Anti-GPR89A Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-137EA 619 EUR
PRSI48-137
Anti-GPR89A Rabbit Polyclonal Antibody
Antikörper
GPR89/SH120 is an Orphan-A GPCR with an unknown ligand.

GPR89A antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR89A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Hamster, Horse, Human, Monkey, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR