Anti-GPR32 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-134EA 619 EUR
PRSI48-134
Anti-GPR32 Rabbit Polyclonal Antibody
Antikörper
GPR32 an Orphan-A GPCR with an unknown ligand.

GPR32 antibody can be used in immunohistochemistry starting at 2.6 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR