Anti-ADAMTS1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-133EA 480 EUR
PRSI48-133
Anti-ADAMTS1 Rabbit Polyclonal Antibody
Antikörper
ADAMTS1 is a Metallopeptidase M12B type protease.

ADAMTS1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 40 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ADAMTS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey, Orangutan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR