Anti-MCHR1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-128EA 619 EUR
PRSI48-128
Anti-MCHR1 Rabbit Polyclonal Antibody
Antikörper
MCHR1 is a Melanin-Concentrating Hormone Receptor that binds MCH to regulate nutritional homeostasis, including feeding and body adiposity. The receptor also functions in sensory perception and integration of smell and gustatory inputs, as well as in emotion and memory.

MCHR1 antibody can be used in ELISA, and immunohistochemistry starting at 15 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MCHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Bovine, Chimpanzee, Dog, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR