Anti-ACAN Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-127EA 480 EUR
PRSI48-127
Anti-ACAN Rabbit Polyclonal Antibody
Antikörper
Aggrecan Core Protein ESTs have been isolated from lung, uterus, brain, colon, bone, adrenal, stomach, breast, liver/spleen, fetal lung, fetal testis, and fetal B-cell libraries.

Aggrecan 1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 1:100, and flow cytometry.

Unpurified

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ACAN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Hamster, Human, Monkey, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR