Anti-MMP13 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-125EA 480 EUR
PRSI48-125
Anti-MMP13 Rabbit Polyclonal Antibody
Antikörper
MMP13 is a Metallopeptidase M10A type protease. Metalloproteinase 13/Collagenase 3 (MMP13) was originally identified in breast carcinoma and has since been documented in a variety of tumors including chondrosarcomas, basal cell carcinoma, and malignant melanoma. In addition it has been detected in inflammatory diseases such as atherosclerosis, in wound healing, and in destructive joint diseases such OA and RA. MMP13 degrades the native helix of several types of fibrillar collagen, and extracellular matrix proteins in vitro.

MMP13 antibody can be used in Western Blot starting at 1 - 10 μg/mL, immunohistochemistry starting at 5 μg/mL, immunocytochemistry starting at 0.1 - 1 μg/mL, and immunoprecipitation starting at 1 - 10 μg/mL

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MMP13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR