Anti-GPR15 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-120EA 480 EUR
PRSI48-120
Anti-GPR15 Rabbit Polyclonal Antibody
G protein-coupled receptor GPR15 is an Orphan-A GPCR with an unknown ligand. Coreceptor usage of primary human immunodeficiency virus type 1 (HIV-1) isolates varies according to biological phenotype. The chemokine receptors CCR5 and CXCR4 are the major coreceptors that, together with CD4, govern HIV-1 entry into cells. Some HIV-1, HIV-2, and SIV isolates of different genetic subtypes and biological phenotypes use BOB/GPR15 for productive infection, suggesting that this cofactor may play a role in HIV-1 pathogenesis and transmission.

GPR15 antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Chimpanzee, Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR