Anti-CCR3 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-113EA 480 EUR
PRSI48-113
Anti-CCR3 Rabbit Polyclonal Antibody
Antikörper
CCR3, a Chemokine Receptor, was cloned from human eosinophils (Daugherty et al., 1996). This receptor binds specifically the chemokine eotaxin and activates eosinophil leukocytes. CCR3 also responds to a variety of beta-chemokines (CC motif) including RANTES, monocyte chemotactic proteins, and mucosae-associated epithelial chemokine (MEC). CCR3 is a key mediator of allergic reactions; activation of this receptor stimulates the production of interleukins required for activation of basophils and eosinophils (Sallusto et al., 1997). CCR3 plays a role in the progression and pathogenesis of inflammatory diseases such as asthma, multiple sclerosis, and Alzheimer's disease. This receptor also facilitates HIV-1 penetration in heart and brain.

CCR3 antibody can be used in ELISA, and immunohistochemistry starting at 20 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CCR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR